Learn More
MSC GF Gage Truncated Taperlock Thread Gage, 3/4-16 UNF, No Go, Class 3A, 3 in. Handle, 1 ea.
Catalog No. 5000514803
Encompass
The GF Gage Truncated Taperlock Thread Gage is a precision instrument designed for verifying the accuracy of 3/4-16 UNF threads. This No Go gage is manufactured to Class 3A specifications to ensure strict tolerance compliance in industrial and manufacturing environments.
  • Truncated taperlock design for precise thread measurement
  • Configured for 3/4-16 UNF thread size
  • Class 3A classification for high-precision verification
  • Includes a 3 in. handle for secure and comfortable operation
  • Constructed from steel for durability and longevity
Catalog No. 50-005-14803 Supplier MSC Supplier No. 55331532
The GF Gage Truncated Taperlock Thread Gage is a precision instrument designed for verifying the accuracy of 3/4-16 UNF threads. This No Go gage is manufactured to Class 3A specifications to ensure strict tolerance compliance in industrial and manufacturing environments.
  • Truncated taperlock design for precise thread measurement
  • Configured for 3/4-16 UNF thread size
  • Class 3A classification for high-precision verification
  • Includes a 3 in. handle for secure and comfortable operation
  • Constructed from steel for durability and longevity