Learn More
MSC GF Gage Truncated Taperlock Thread Gage, #6-32 UNC, No Go, Class 3A, 1 ea
Catalog No. 5000514727
Encompass
This truncated taperlock thread gage is designed for precise inspection of #6-32 UNC internal threads. Manufactured from durable steel, this gage is configured as a no go gauge with a 3A classification to ensure accuracy and compliance with thread specifications in high-precision engineering and manufacturing applications.
  • Truncated taperlock thread gage design
  • No go classification for thread fit verification
  • Constructed from steel for durability
  • Designed for #6-32 UNC thread sizes
  • Class 3A classification
Catalog No. 50-005-14727 Supplier MSC Supplier No. 55330930
This truncated taperlock thread gage is designed for precise inspection of #6-32 UNC internal threads. Manufactured from durable steel, this gage is configured as a no go gauge with a 3A classification to ensure accuracy and compliance with thread specifications in high-precision engineering and manufacturing applications.
  • Truncated taperlock thread gage design
  • No go classification for thread fit verification
  • Constructed from steel for durability
  • Designed for #6-32 UNC thread sizes
  • Class 3A classification