Learn More
MSC GF Gage Truncated Taperlock Thread Gage, Steel, #12-24, Go, 3A, 1 ea
Catalog No. 5000511528
Encompass
This truncated taperlock thread gage is a precision instrument designed for accurate thread verification. Engineered for reliability, this gage is suitable for inspecting #12-24 threads in various metalworking applications.
  • Constructed from durable steel for long-lasting wear resistance
  • Features a truncated taperlock design for consistent measurements
  • Designed for go thread inspection applications
  • Thread size of #12-24
  • Manufactured to 3A classification standards
Catalog No. 50-005-11528 Supplier MSC Supplier No. 55328736
This truncated taperlock thread gage is a precision instrument designed for accurate thread verification. Engineered for reliability, this gage is suitable for inspecting #12-24 threads in various metalworking applications.
  • Constructed from durable steel for long-lasting wear resistance
  • Features a truncated taperlock design for consistent measurements
  • Designed for go thread inspection applications
  • Thread size of #12-24
  • Manufactured to 3A classification standards