Learn More
MSC GF Gage Truncated Taperlock Thread Plug Gage, Steel, 9/16-18, Go, 1 ea.
Catalog No. 5000514526
Encompass
This truncated taperlock thread gage is a precision measuring instrument designed for verifying the accuracy of 9/16-18 threaded parts. Engineered for reliability in industrial inspection environments, this go plug gage ensures that internal threads meet established specifications.
  • Manufactured from durable steel
  • Designed for 9/16-18 thread size applications
  • Configured as a go gage for thread setting
  • Includes a 3 in. handle
  • Made in the United States
Catalog No. 50-005-14526 Supplier MSC Supplier No. 55329098
This truncated taperlock thread gage is a precision measuring instrument designed for verifying the accuracy of 9/16-18 threaded parts. Engineered for reliability in industrial inspection environments, this go plug gage ensures that internal threads meet established specifications.
  • Manufactured from durable steel
  • Designed for 9/16-18 thread size applications
  • Configured as a go gage for thread setting
  • Includes a 3 in. handle
  • Made in the United States