Learn More
Aviva Systems Biology Corp GFM2 Antibody - C-terminal region (ARP60546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFM2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GFM2. Alternate names: EFG2, MRRF2, MST027, RRF2, RRF2mt, hEFG2, EF-G2mt, mEF-G 2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-287-080