Learn More
Santa Cruz Biotechnology GFP (1-238) 10 UG/0.1 ML
Supplier:  Santa Cruz Biotechnology SC4304 WB
Encompass
Full length (amino acids 1-238) GFP of Aequorea victoria origin. produced in E. coli as 53 kDa tagged fusion protein. provided as 10 ug protein in 0.1 ml SDS-PAGE loading buffer. WB control for sc-5384, sc-5385, sc-8334 and sc-9996.
Catalog No. NC0887469