Learn More
Santa Cruz Biotechnology GFP (B-2)
Supplier:  Santa Cruz Biotechnology SC9996
Encompass
GFP (B-2), mouse monoclonal IgG2a; 200microgram/ml, raised against amino acids 1-238 representing full length GFP (green fluorescent protein) of Aequorea victoria origin, reactive with GFP and GFP mutant fusion proteins
Catalog No. NC9830411