Learn More
Proteintech Group Inc GFP-Booster Alexa Fluor 647
SDP
Catalog No. 502258316 Shop All Proteintech Group Inc Products
Encompass
anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy.
This item is not returnable. View return policy
Catalog No. 50-225-8316 Supplier Proteintech Group Inc Supplier No. GB2AF64750
anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy.