Learn More
Proteintech Group Inc GFP-Booster ATTO488
SDP
Supplier:  Proteintech Group Inc GBA488100
Encompass
anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy.
Catalog No. 50-225-8318