Learn More
Enzo Life Sciences GFP-CERTIFIED FLUOFORTE Reagent (Non-AM) (50µg)
SDP
Catalog No. 502012646
Encompass_Preferred
The GFP-CERTIFIED FLUOFORTE Reagent (Non-AM) is the most sensitive fluorescent calcium indicator relative to other commercially available red fluorescent dyes. It can be used for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.
Catalog No. 50-201-2646 Supplier Enzo Life Sciences Supplier No. ENZ525550250
The GFP-CERTIFIED FLUOFORTE Reagent (Non-AM) is the most sensitive fluorescent calcium indicator relative to other commercially available red fluorescent dyes. It can be used for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Esterase hydrolysis is not required for live cell analysis via flow cytometry or fluorescence microscopy. Purity: ≥95% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.