Learn More
Enzo Life Sciences GFP-CERTIFIED FLUOFORTE Reagent (50µg)
SDP
Supplier:  Enzo Life Sciences ENZ520165C50
Encompass_Preferred
The GFP-CERTIFIED FLUOFORTE Reagent is the brightest and most sensitive fluorescent calcium indictor relative to other commercially available red fluorescent dyes with maximum excitation at ~530nm and maximum emission at ~555nm. This product is used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C. Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection, HTS .
Catalog No. 50-201-2611