Learn More
Enzo Life Sciences GFP-CERTIFIED Apoptosis/Necrosis detection kit (1x25assays)
SDP
Catalog No. NC0492476
Encompass_Preferred
Multiplex assay that distinguishes between  healthy, early apoptotic, late apoptotic and necrotic cells, compatible with GFP and other green fluorescent probes. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection. Contents: Apoptosis Detection Reagent (Annexin V-EnzoGold). Necrosis Detection Reagent. Apoptosis Inducer (Staurosporine). Binding Buffer (10X)
Catalog No. NC0492476 Supplier Enzo Life Sciences Supplier No. ENZ5100225
Multiplex assay that distinguishes between  healthy, early apoptotic, late apoptotic and necrotic cells, compatible with GFP and other green fluorescent probes. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection. Contents: Apoptosis Detection Reagent (Annexin V-EnzoGold). Necrosis Detection Reagent. Apoptosis Inducer (Staurosporine). Binding Buffer (10X)