Learn More
Gentarget GFP (CMV, Zeo) lentiviral particles in PBS
Catalog No. NC3364808
Encompass
Pre-made RFP lentiviral particles expressing codon optimized Green Fluorescent Protein (GFP) marker under suCMV promoter. A Zeocin antibiotic selection under Rsv promoter allows to select transduced fluorescent positive cells. The following scheme showed the lentivector expression cassette:Amount: 1 x 108 IFU/ml x 200ul in PBS. (This is the concentrated lentivirus with higher titer)
Catalog No. NC3364808 Supplier Gentarget Supplier No. LVP1231PBS
Pre-made RFP lentiviral particles expressing codon optimized Green Fluorescent Protein (GFP) marker under suCMV promoter. A Zeocin antibiotic selection under Rsv promoter allows to select transduced fluorescent positive cells. The following scheme showed the lentivector expression cassette:Amount: 1 x 108 IFU/ml x 200ul in PBS. (This is the concentrated lentivirus with higher titer)