Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFP-EIF2A

Catalog No. PV4809
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4809 20 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4809 Supplier Invitrogen™ Supplier No. PV4809
  • $1,202.00
    $1,177.65 / Each of 1

    Save $24.35

    2% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GFP-EIF2A

EIF2A has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2).

For use in combination with the LanthaScreen™ Tb-anti-eIF2alpha [pSer52] Antibody (PV4810) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage GFP-EIF2alpha protein substrate in 9 mM HEPES pH 7.5, 123 mM NaCl, 2.43 mM KCl, 10% Glycerol, 2mM DTT . Store at -80 °C.
Detection Method Fluorescence
Form Liquid
Quantity 20 nmol
Substrate EIF2A
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 9833
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 69100 Da
Shipping Condition Dry Ice
Target Entry EIF2AK1, EIF2AK2, EIF2AK3
Validated Kinases EIF2AK2, EIF2AK3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.