Learn More
Angio Proteomie GFP EXPRESSING HUMAN DERMAL MI
SDP
Supplier:  Angio Proteomie CAP0005GFP
Encompass
GFP-HDMVECs were selected from HDMVECs (cAP-0004) resistant to puromycin after infected with GFP expressing lentiviral particles. The cells are shipped in frozen vials (the cells are provided @ passage 3). .
Catalog No. NC2315759