Learn More
Gentarget GFP-Histone2B (Neo) Fusion, Concentrated Lentivirus, 200ul/vial (1×108 IFU/ml), (provided in PBS with 60ug/ml Polybrene (10x).
Catalog No. NC3883534
Encompass
Pre-made lentivirus express a fusion target of (GFP-human Histone 2B). The human Histone 2B (H2B) was fusioned at its N-terminal with GFP, expressed under a constitutive suCMV promoter, containing a Neomycin antibiotic marker under the RSV promoter.Fluorescent labeled H2B allowed high-resolution imaging of both mitotic chromosomes and interphase chromatin, and the latter revealed various chromatin condensation states in live cells. It can directly observe the Double Minute Chromosomes (DMs) in living cells without compromising nuclear and chromosomal structures.
Catalog No. NC3883534 Supplier Gentarget Supplier No. LVP444GNPBS
Pre-made lentivirus express a fusion target of (GFP-human Histone 2B). The human Histone 2B (H2B) was fusioned at its N-terminal with GFP, expressed under a constitutive suCMV promoter, containing a Neomycin antibiotic marker under the RSV promoter.Fluorescent labeled H2B allowed high-resolution imaging of both mitotic chromosomes and interphase chromatin, and the latter revealed various chromatin condensation states in live cells. It can directly observe the Double Minute Chromosomes (DMs) in living cells without compromising nuclear and chromosomal structures.