Learn More
Enzo Life Sciences Green Fluorescent Protein monoclonal antibody (1E4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADISAB500E
Encompass_Preferred
Clone: 1E4. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant GFP protein (Aequorea victoria). Species reactivity: Species independent. Applications: ICC, IHC, IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7397