Learn More
Enzo Life Sciences Green fluorescent protein monoclonal antibody (B34) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS1410200
Encompass_Preferred
Clone: B34. Isotype: IgG1. Immunogen: Purified wild type GFP. Specificity: Recognizes GFP and GFP variants including enhance/mutant GFP. Applications: WB. Formulation: Liquid. In PBS with 0.03% thimerosal.
Catalog No. 50-201-2702