Learn More
Abcam Recombinant A. victoria GFP protein, 100UG
Supplier:  Abcam AB84191100UG
Encompass_Preferred
Recombinant A. victoria GFP protein is a Aequorea victoria Full Length protein, in the 1 to 238 aa range with >95% purity, < 1 EU/µg endotoxin level and suitable for western blot, Functional studies and SDS-PAGE. The predicted molecular weight of ab84191 recombinant protein is 26.8 kDa. - Save time and ensure accurate results - use our recombinant GFP protein as a control - Available in different sizes to fit your experimental needs
Protein/ Peptide/ Enzyme Synonyms: Green fluorescent protein, GFP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0403232