Learn More
Cell Biolabs Inc GFP Quantitation Kit (Fluorometric), 100 assays
Catalog No. NC9827506
Encompass_Preferred
Most imaging studies of rGFP are qualitative. Quantitative analysis of rGFP level in cells by FACS is time consuming and expensive. Cell Biolabs’ GFP Quantitation Kit provides a convenient plate-based alternative to measuring GFP fluorescence by FACS analysis. The quantity of rGFP in sample is determined by comparing its fluorescence reading with that of known recombinant GFP standard curve. The kit has detection sensitivity limit of 100 ng/mL. A proprietary GFP quench solution is also included for determining autofluorescence of cell or tissue sample. Each kit provides sufficient reagents to perform up to 100 assays including standard curve and GFP samples.
Catalog No. NC9827506 Supplier Cell Biolabs Inc Supplier No. AKR120
Most imaging studies of rGFP are qualitative. Quantitative analysis of rGFP level in cells by FACS is time consuming and expensive. Cell Biolabs’ GFP Quantitation Kit provides a convenient plate-based alternative to measuring GFP fluorescence by FACS analysis. The quantity of rGFP in sample is determined by comparing its fluorescence reading with that of known recombinant GFP standard curve. The kit has detection sensitivity limit of 100 ng/mL. A proprietary GFP quench solution is also included for determining autofluorescence of cell or tissue sample. Each kit provides sufficient reagents to perform up to 100 assays including standard curve and GFP samples.