Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GFP-STAT3

Catalog No. PV5212
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5212 10 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5212 Supplier Thermo Scientific™ Supplier No. PV5212
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STAT3 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

STAT3 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen™ Tb-anti-STAT3 [pTyr705] Antibody (Cat. No. PV4846) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage GFP-STAT3 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT

Store at -80°C.
Detection Method Fluorescence
Quantity 10 nmol
Substrate STAT3
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Tyrosine Kinases
Label or Dye GFP (EmGFP)
Molecular Weight (g/mol) 145,200 Da
Shipping Condition Dry Ice
Validated Kinases JAK1, JAK2, JAK3, TYK2
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.