Learn More
Proteintech Group Inc GFP-Trap Agarose
SDP
Supplier:  Proteintech Group Inc GTA100
Encompass
GFP-Trap Agarose is an affinity resin for IP of GFP-fusion proteins. It consists of a GFP Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8329