Learn More
Proteintech Group Inc GFP-Trap Magnetic Agarose
SDP
Catalog No. 502258340 Shop All Proteintech Group Inc Products
Encompass
The ChromoTek GFP-Trap Magnetic Agarose are affinity beads for IP of GFP-fusion proteins. It comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8340 Supplier Proteintech Group Inc Supplier No. GTMA10
 Restricted Item
Add to Cart
Add to Cart
The ChromoTek GFP-Trap Magnetic Agarose are affinity beads for IP of GFP-fusion proteins. It comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads.