Learn More
Proteintech Group Inc GFP-Trap Magnetic Agarose, kit
SDP
Supplier:  Proteintech Group Inc GTMAK20
Encompass
The ChromoTek GFP-Trap Magnetic Agarose are affinity beads for IP of GFP-fusion proteins. It comprises a GFP Nanobody/ VHH coupled to magnetic agarose beads.
Catalog No. 50-225-8345