Learn More
Proteintech Group Inc GFP-Trap Magnetic Particles M-270
SDP
Supplier:  Proteintech Group Inc GTD100
Encompass
GFP-Trap Magnetic Particles M-270 for immunoprecipitation (IP) of GFP-tagged proteins. GFP-Trap Magnetic Particles M-270 is highly recommended, when very large proteins/complexes are investigated, and magnetic separation is needed for IP. It consists of a GFP VHH/ Nanobody coupled to Magnetic Particles M-270.
Catalog No. 50-225-8336