Learn More
Aviva Systems Biology Corp GFPT2 Antibody - N-terminal region (ARP42214_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFPT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GFPT2. Alternate names: FLJ10380, GFAT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-984