Learn More
Bulldog Bio Inc GFPTRAP MAG AGAROSE 2.5ML
SDP
Catalog No. 50999504
Encompass
Reagent; Molecular Biology; GFP-Trap, Coupled to Magnetic Agarose Beads, for Immuno-Pulldown of GFP Fusions, 100 Reactions, 2.5ml
Catalog No. 50-999-504 Supplier Bulldog Bio Inc Supplier No. GTMA100
Reagent; Molecular Biology; GFP-Trap, Coupled to Magnetic Agarose Beads, for Immuno-Pulldown of GFP Fusions, 100 Reactions, 2.5ml