Learn More
Aviva Systems Biology Corp GFRA2 Antibody - C-terminal region (ARP54575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFRA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GFRA2. Alternate names: GDNFRB, NRTNR-ALPHA, NTNRA, RETL2, TRNR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53741