Learn More
Aviva Systems Biology Corp GFRA4 Antibody - C-terminal region (ARP61586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFRA4. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Human
Catalog No. 50-287-412