Learn More
Genscript Corporation GFRAL/GFR alpha-likeBiotin
Catalog No. 5000058023
Encompass
GFR alpha -like (GDNF receptor-alpha-like) is a distant member of the GDNFR family of proteins Mature human GFR alpha-like is a 376 amino acid (aa) type I transmembrane protein It contains a 333 aa extracellular domain a 20 aa transmembrane domain and a 23 aa cytoplasmic domain GFRAL is a brainstem-restricted receptor for GDF15 which regulates food intake energy expenditure and body weight in response to metabolic and toxin-induced stresses
Catalog No. 50-000-58023 Supplier Genscript Corporation Supplier No. Z04471100
GFR alpha -like (GDNF receptor-alpha-like) is a distant member of the GDNFR family of proteins Mature human GFR alpha-like is a 376 amino acid (aa) type I transmembrane protein It contains a 333 aa extracellular domain a 20 aa transmembrane domain and a 23 aa cytoplasmic domain GFRAL is a brainstem-restricted receptor for GDF15 which regulates food intake energy expenditure and body weight in response to metabolic and toxin-induced stresses