Learn More
Medchemexpress LLC GFRAL Human Biotin 50ug | 50ug
SDP
Catalog No. 5000193869
Encompass_Preferred
The GFRAL protein is a brainstem-restricted receptor for GDF15 that regulates food intake energy expenditure and body weight in response to metabolic and toxin-induced stress GFRAL binds to its ligand GDF15 interacts with RET and activates the MAPK and AKT signaling pathways GFRAL Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived GFRAL protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93869 Supplier Medchemexpress LLC Supplier No. HYP7813550UG
The GFRAL protein is a brainstem-restricted receptor for GDF15 that regulates food intake energy expenditure and body weight in response to metabolic and toxin-induced stress GFRAL binds to its ligand GDF15 interacts with RET and activates the MAPK and AKT signaling pathways GFRAL Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived GFRAL protein expressed by HEK293 with C-Avi C-His labeled tag