Learn More
Medchemexpress LLC GFRAL Human His-SU 100ug
SDP
Catalog No. 5000196311
Encompass_Preferred
Recombinant human GFRAL protein (residues S19-E351, accession Q6UXV0) supplied as an N-terminal His-SUMO fusion expressed in E. coli. Purified protein (typical purity >95% by SDS-PAGE) provided in a 100 μg format for use in biochemical and cell-based studies of GFRAL-GDF15 signaling and receptor interactions.
  • N-terminal His-SUMO fusion for affinity purification.
  • Sequence spans residues S19-E351 of the human protein.
  • Expressed in E. coli and purified to >95% purity by SDS-PAGE.
  • Supplied as a 100 μg format suitable for assay use.
  • Intended for binding assays, antibody characterization, and receptor interaction studies.
  • Batch-specific certificate of analysis available with accession Q6UXV0 sequence reference.
Catalog No. 50-001-96311 Supplier Medchemexpress LLC Supplier No. HYP71580100UG
Recombinant human GFRAL protein (residues S19-E351, accession Q6UXV0) supplied as an N-terminal His-SUMO fusion expressed in E. coli. Purified protein (typical purity >95% by SDS-PAGE) provided in a 100 μg format for use in biochemical and cell-based studies of GFRAL-GDF15 signaling and receptor interactions.
  • N-terminal His-SUMO fusion for affinity purification.
  • Sequence spans residues S19-E351 of the human protein.
  • Expressed in E. coli and purified to >95% purity by SDS-PAGE.
  • Supplied as a 100 μg format suitable for assay use.
  • Intended for binding assays, antibody characterization, and receptor interaction studies.
  • Batch-specific certificate of analysis available with accession Q6UXV0 sequence reference.