Recombinant human GFRAL protein comprising residues S19-E351 with an N-terminal His-SUMO fusion, expressed in E. coli and supplied as purified research-grade material. It is intended for biochemical and cell biology studies that require the extracellular/receptor fragment of GFRAL, and includes recommended storage and handling instructions to preserve integrity and activity.
- Sequence corresponds to UniProt Q6UXV0, residues S19-E351.
- N-terminal His and SUMO fusion tags for purification and stability.
- Expressed in E. coli and purified to 93.0% as determined by reducing SDS-PAGE.
- Typical supplied quantity: 50 μg (lyophilized).
- Storage: store at -20°C for long term; after reconstitution stable at 4°C for 1 week; aliquot and freeze at -20°C or -80°C for extended storage.
- Suitable for biochemical assays, binding studies, and cell-based experiments.
- Provided with a certificate of analysis for lot-specific quality assurance.