Learn More
ACROBiosystems ACROBiosystems Human GFR alpha-like Protein, His Tag
Supplier: ACROBiosystems GFAH52H3100UG
Recombinant Protein;100UG;Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession # Q6UXV0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.