Learn More
ACROBiosystems Recombinant Protein;Human GFR alpha-1 Protein;HEK293;1MG;GF1,GFRA1,GDNFRA,RETL1,TRNR1,GDNFR,GFR-ALPHA-1,RET1L,GFRA-1,RETL-1,TRNR-1
Catalog No. 501657833
Encompass_Preferred
Recombinant Protein;Human GFR alpha-1 Protein;1MG;Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 424 (Accession # NP_665736.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.5 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7833 Supplier ACROBiosystems Supplier No. GF1H52241MG
Recombinant Protein;Human GFR alpha-1 Protein;1MG;Human GFR alpha-1, His Tag (GF1-H5224) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ser 424 (Accession # NP_665736.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 45.5 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.