Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ QuickSlide™ GramPRO 1 Automated Gram Stain Instrument Reagent Kit
SDP

Catalog No. 23001200 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type:
Reagent Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-001-200 Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-001-200 Supplier Hardy Diagnostics™ Supplier No. GP1SK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient Reagent kit. Hardy Diagnostics™ QuickSlide™ GramPRO 1 Automated Gram Stain Instrument Reagent Kit includes Crystal Violet, Safranin, Acetone Alcohol, and Grams Iodine

  • Extended shelf life of two years (unopened)
  • Special formulated pack - 110 tests/ Pk. in one self contained package
  • Easy to read labels on tubing for ease of operation

Recommended Storage

Keep container tightly closed. Store in a cool, dry place.

Specifications

Product Type Reagent Kit
Color Clear Bottles
For Use With (Equipment) AGS-00700, AGS-1000, AGS-SC-4000, AGS-TK-3000
Includes Crystal Violet, Safranin, Acetone Alcohol, and Grams Iodine. 110 tests per pack in one self-contained package
Weight (English) 7 lb.
Weight (Metric) 3.17 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.