Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ QuickSlide™ HemaPRO Automated Hematology Stain Instrument
SDP

Catalog No. 2300111 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type:
Automated Hematol.Stain Instr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-001-11 Automated Hematol.Stain Instr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-001-11 Supplier Hardy Diagnostics™ Supplier No. HP1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Represents a significant advancement in automated slide staining. Hardy Diagnostics™ QuickSlide™ HemaPRO Automated Hematology Stain Instrument features soft agitation action after staining

  • Operator prompt LCD display
  • Soft touch color-coded keys
  • Adjustable stain and buffer duration
  • Adjustable fill level
  • “Cycles Remaining” display - computer monitors reagent and prompts when to reorder
  • Injection molded case with built-in reagent platform
  • Extremely durable
  • Color-coded reagent tubing for ease of identification
  • Special formulated reagent kit -125 tests/ kit in one self-contained package
  • Used as primary stainer used by small hospitals, physicians labs, stat labs, or as a stat/backup instrument in larger labs
  • Uses modified Wrights-Giemsa stain specially formulated for the brightest colors
  • Features soft agitation action after staining to ensure clean slide
  • Needs no daily maintenance and requires a tubing change once or twice annually
  • Uses fresh stain on every slide to reduce the risk of cross-contamination

Recommended Storage

Wright Giemsa Stain -Keep container tightly closed. Store away from all sources of ignition. Store in a cool, dry, well-ventilated place.

Specifications

Product Type Automated Hematol.Stain Instr.
Color Black
For Use With (Application) QS-002 and QS-29000
Includes Power cord, User manual
Temperature Range (Metric) 16°C to 32°C
Weight (English) 17 lb.
Weight (Metric) 7.7 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.