Learn More
ProSci GGA3 Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 23163 |
| Accession Number | NP_619525.1, NP_054720.1 |
| Common Name | golgi-associated, gamma adaptin ear containing, ARF binding protein 3 |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | GGA3 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | GGA3 |
| Quantity | 0.1 mg |
| Sequence | GGA3 peptide corresponds to 12 amino acids of GGA3. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.