Learn More
Haematologic Technologies Inc GGACK 5MG
Catalog No. 50883357
Encompass_Preferred
Inhibitor, Glu-Gly-Arg Chloromethylketone (GGACK), 5 milligrams, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, Supplied lyophilized, store at 4 degrees Celsius.
Catalog No. 50-883-357 Supplier Haematologic Technologies Inc Supplier No. EGRCK015MG
Inhibitor, Glu-Gly-Arg Chloromethylketone (GGACK), 5 milligrams, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, Supplied lyophilized, store at 4 degrees Celsius.