Learn More
Cayman Chemical GGACKhydrochlorIde 10mg
SDP
Catalog No. 502950265
Encompass_Preferred
A uPA and factor Xa inhibitor (Kis = 1.8 and 8.7 µM, respectively)
Catalog No. 50-295-0265 Supplier Cayman Chemical Supplier No. 3461810 MG
A uPA and factor Xa inhibitor (Kis = 1.8 and 8.7 µM, respectively)