Learn More
Aviva Systems Biology Corp GGCX Antibody - middle region (ARP44340_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44340_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GGCX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GGCX. Alternate names: FLJ26629, VKCFD1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58785