Learn More
Adam Equipment GGF 300aM - Gladiator Approved Washdown Scales - Capacity: 300lb / 150kg, Readability: 0.05lb / 0.02kg
Supplier:  Adam Equipment GGF 300AM
Encompass_Preferred
For professionals in food, pharmaceutical, and industrial sectors, the Gladiator M NTEP-approved waterproof scale delivers precise, reliable performance in washdown environments. With an IP68 rating and durable stainless-steel construction, it offers superior protection against water and dust ensuring easy cleaning and long-term durability. Designed for efficiency, it features a large backlit display with color-changing checkweighing indicators for quick visual feedback, along with a tactile keypad with large, color-coded keys for fast operation. Built for trade use, it’s ideal when weight determines pricing. Heavy-duty rubber feet provide stability, while the sealed keypad protects against spills. The RS-232 interface enables seamless data transfer, and the rechargeable battery supports portability with auto power-off for energy savings. External calibration, hold function, parts counting, digital filtering, and preset tare ensure accurate, and versatile weighing performance.
Catalog No. NC3964915