Learn More
Adam Equipment Adam Equipment Gladiator Washdown Scale GGF 165aH 165lb / 75kg capacity x 0.005lb / 2g readability
Catalog No. NC2612394
Encompass_Preferred
Adam Equipment GGF Gladiator Washdown Scales feature a combination of durability and performance, Gladiator scales accomplish washdown weighing tasks in demanding food, pharmaceutical and industrial settings. With an IP68 rating for washdown capabilities, Gladiator features easy-to-clean stainless steel construction that is ideal for wet and dusty environments. Connectors on the bottom are IP68-rated for thorough protection. Gladiator can expedite checkweighing tasks, with a large, backlit display that changes color to show if the weight is over, under or within the set limit. The keypad is designed for maximum efficiency, with large, color-coded tactile keys for fast recognition and easy operation. Rechargeable battery included for operation almost anywhere.
Catalog No. NC2612394 Supplier Adam Equipment Supplier No. GGF 165AH
Adam Equipment GGF Gladiator Washdown Scales feature a combination of durability and performance, Gladiator scales accomplish washdown weighing tasks in demanding food, pharmaceutical and industrial settings. With an IP68 rating for washdown capabilities, Gladiator features easy-to-clean stainless steel construction that is ideal for wet and dusty environments. Connectors on the bottom are IP68-rated for thorough protection. Gladiator can expedite checkweighing tasks, with a large, backlit display that changes color to show if the weight is over, under or within the set limit. The keypad is designed for maximum efficiency, with large, color-coded tactile keys for fast recognition and easy operation. Rechargeable battery included for operation almost anywhere.