Learn More
Medchemexpress LLC Ggfg-amide-glycol-amide-exatecan | 2866301-18-8 | 98.7% | 870.88 g/mol | C43H47FN8O11 | 10 MG

Supplier: Medchemexpress LLC HY15360010MG
GGFG-amide-glycol-amide-Exatecan is an Exatecan-derived small-molecule intermediate used for research and for assembling antibody-drug conjugates (ADCs). It functions as a DNA topoisomerase I inhibitor derivative and is supplied as a high-purity solid for laboratory use.
- Intermediate for antibody-drug conjugate synthesis.
- Cas number 2866301-18-8.
- Molecular formula C43H47FN8O11; molecular weight 870.88 g/mol.
- Reported purity 98.7%.
- Appearance: off-white to light yellow solid.
- Solubility: DMSO 100 mg/mL (ultrasonic assistance recommended); hygroscopic effects reported.
- Storage: sealed, away from moisture and light; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.