Learn More
Medchemexpress LLC Gggyk-biotin | >99.75% | 706.81 | 5 MG
SDP
Catalog No. 5000425538
Encompass_Preferred
Gggyk-biotin is a synthetic peptide that serves as a substrate for Sortase A, an enzyme involved in bacterial surface protein anchoring. This peptide is specifically designed to facilitate studies on the substrate specificity of Sortase A, allowing researchers to explore and develop variants of the enzyme with altered substrate preferences.
  • Acts as a substrate for Sortase A.
  • Useful for studying Sortase A's substrate specificity.
  • Can be employed in the development of Sortase A variants with distinct substrate specificities.
  • Intended for research purposes and not for human use.
Catalog No. 50-004-25538 Supplier Medchemexpress LLC Supplier No. HYP106205MG
Gggyk-biotin is a synthetic peptide that serves as a substrate for Sortase A, an enzyme involved in bacterial surface protein anchoring. This peptide is specifically designed to facilitate studies on the substrate specificity of Sortase A, allowing researchers to explore and develop variants of the enzyme with altered substrate preferences.
  • Acts as a substrate for Sortase A.
  • Useful for studying Sortase A's substrate specificity.
  • Can be employed in the development of Sortase A variants with distinct substrate specificities.
  • Intended for research purposes and not for human use.