Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GGGYK-Biotin | 99.8% | 706.81 | 10 MG
SDP

Catalog No. 5000425603
Encompass_Preferred

GGGYK-Biotin is a substrate peptide designed to study the substrate specificity of Sortase A. It can be used to develop Sortase A variants with different substrate specificities.

  • Purity of 99.75%
  • Molecular weight of 706.81
  • Appears as a white to off-white solid
  • Sequence: Gly-Gly-Gly-Tyr-Lys-Biotin
  • Formula: C31H46N8O9S

Catalog No. 50-004-25603 Supplier Medchemexpress LLC Supplier No. HYP1062010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GGGYK-Biotin is a substrate peptide designed to study the substrate specificity of Sortase A. It can be used to develop Sortase A variants with different substrate specificities.

  • Purity of 99.75%
  • Molecular weight of 706.81
  • Appears as a white to off-white solid
  • Sequence: Gly-Gly-Gly-Tyr-Lys-Biotin
  • Formula: C31H46N8O9S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.