Learn More
Adam Equipment GGS 16a - Gladiator Washdown Scales - Capacity: 16lb / 8kg, Readability: 0.001lb / 0.5g
Catalog No. NC3964917
Encompass_Preferred
Gladiator Washdown Scales deliver precision and durability for food, pharmaceutical, and industrial environments. With an IP68 rating and stainless steel construction, they perform reliably in wet and dusty conditions while remaining easy to clean. The large backlit display changes color to indicate over, under, or within limits, simplifying checkweighing, while the sealed keypad with color-coded keys ensures easy operation.Available in multiple platform sizes and capacities, Gladiator scales support weighing, parts counting, percentage weighing, and dynamic weighing. An RS-232 interface enables seamless data transfer to PCs and printers. Heavy-duty rubber feet provide stability, and a rechargeable battery with auto power-off enhances portability and efficiency.Backed by a 5-year warranty, Gladiator scales offer dependable performance and accuracy for demanding applications.
Catalog No. NC3964917 Supplier Adam Equipment Supplier No. GGS 16A
Gladiator Washdown Scales deliver precision and durability for food, pharmaceutical, and industrial environments. With an IP68 rating and stainless steel construction, they perform reliably in wet and dusty conditions while remaining easy to clean. The large backlit display changes color to indicate over, under, or within limits, simplifying checkweighing, while the sealed keypad with color-coded keys ensures easy operation.Available in multiple platform sizes and capacities, Gladiator scales support weighing, parts counting, percentage weighing, and dynamic weighing. An RS-232 interface enables seamless data transfer to PCs and printers. Heavy-duty rubber feet provide stability, and a rechargeable battery with auto power-off enhances portability and efficiency.Backed by a 5-year warranty, Gladiator scales offer dependable performance and accuracy for demanding applications.