Learn More
Adam Equipment Trade Approved Gladiator Washdown Scale GGS30aM 30lb / 15kg capacity x 0.005lb / 2g readability
Supplier:  Adam Equipment GGS 30AM
Encompass_Preferred
Adam Equipment GGS Trade Aproved Gladiator Washdown Scales featuring a combination of durability and performance, Gladiator M NTEP-approved scales perform washdown weighing tasks in demanding food, pharmaceutical and industrial settings. Boasting an IP68 rating for washdown capabilities, Gladiator M features easy-to-clean stainless steel construction that is ideal for wet and dusty environments. Connectors on the bottom are IP68-rated for thorough protection. Designed for maximum efficiency, Gladiator M expedites checkweighing tasks with a large, backlit display that changes color to show if the weight is over, under or within the set limit. The tacile keypad offers large, color-coded keys for fast recognition and easy operation. Suitable for trade use, where a weighing scale is needed to determine a price for goods being bought or sold. Rechargeable battery included for operation almost anywhere.
Catalog No. NC2611838