Learn More
Cayman Chemical GGT Assay Buffer, 25 mL
SDP
Catalog No. 502945376
Encompass_Preferred
GGT assay buffer has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).
Catalog No. 50-294-5376 Supplier Cayman Chemical Supplier No. 40051425 ML
GGT assay buffer has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).