Learn More
Cayman Chemical GGT Positive Control, 1 ea
SDP
Catalog No. 502942463
Encompass_Preferred
GGT positive control has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).
Catalog No. 50-294-2463 Supplier Cayman Chemical Supplier No. 4005181 EA
GGT positive control has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).